Anti-MAP Kinase ERK1/ERK2 (pThr 202 /Tyr 204 ) (12D4) Mouse Monoclonal Antibody
442706-1
- Antibody type:Primary
- Antigen name:MAP Kinase ERK1/ERK2
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Modification:phospho-Thr202/phospho-Tyr204
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 Set
- Pk:1 SET
Type: Primary
Antigen: MAP Kinase ERK1/ERK2 (pThr 202 /Tyr 204 ) (12D4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: