Anti-SLC6A3 Mouse Monoclonal Antibody (DyLight® 350) [clone: mAb16]
Catalog # NOVUNBP2-22164UV
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 6 (neurotransmitter transporter), member 3
- Antigen symbol:Solute carrier family 6, member 3
- Clonality:Monoclonal
- Clone:mAb16
- Conjugation:DyLight® 350
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:6531
- Antigen synonyms:DAT1|PKDYS|DAT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: SLC6A3 (solute carrier family 6 (neurotransmitter transporter), member 3)
Clonality: monoclonal
Clone: mAb16
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat