Anti-RBFOX1 Mouse Monoclonal Antibody [clone: D8H8]
Type: Primary
Antigen: RBFOX1 (RNA binding protein, fox-1 homolog (C. elegans) 1)
Clonality: monoclonal
Clone: D8H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
- Catalog No:
- NOVUNBP2-13169SS
- NOVUNBP2-13169
- Antigen symbol:
- RBFOX1
- RBFOX1
- Antigen name:
- RNA Binding Protein, Fox-1 homolog 1
- RNA Binding Protein, Fox-1 homolog 1
- Antigen synonyms:
- 2BP1|FOX1|HRNBP1|FOX-1|A2BP1
- 2BP1|FOX1|HRNBP1|FOX-1|A2BP1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- D8H8
- D8H8
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 54715
- 54715
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together