Anti-DGAT2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
NOVUNBP1-71701H
- Antibody type:Primary
- Antigen name:Diacylglycerol O-acyltransferase 2
- Antigen symbol:DGAT2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:84649
- Antigen synonyms:HMFN1045|GS1999FULL|ARAT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: DGAT2 (diacylglycerol O-acyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse