Anti-SLC31A2 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-05199B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Copper Transporter 2
- Antigen symbol:CTR2
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:1318
- Antigen synonyms:CTR2|hCTR2|COPT2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SLC31A2 (solute carrier family 31 (copper transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse