Order Entry
Ireland
ContactUsLinkComponent
 
Anti-STRN3 Mouse Monoclonal Antibody [clone: S68]
 
Anti-STRN3 Mouse Monoclonal Antibody [clone: S68]

 

  • Catalog No:
  • NOVUNB110-74572SS
  • NOVUNB110-74572
  • Antigen name:
  • Striatin, Calmodulin Binding Protein 3
  • Striatin, Calmodulin Binding Protein 3
  • Antigen synonyms:
  • STRN3 |Cell-cycle autoantigen SG2NA |SG2NA |S/G2 antigen |GS2NA
  • STRN3 |Cell-cycle autoantigen SG2NA |SG2NA |S/G2 antigen |GS2NA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • S68
  • S68
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 29966
  • 29966
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together