Anti-PLIN5 Rabbit Polyclonal Antibody (DyLight® 550)
NOVUNB110-60509R
- Antibody type:Primary
- Antigen name:Perilipin-5
- Antigen symbol:OXPAT
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:440503
- Antigen synonyms:MLDP|LSDA5|LSDP5|OXPAT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: PLIN5 (perilipin 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: