Anti-NRIP Rabbit Polyclonal Antibody
NOVUNB100-56434
- Antibody type:Primary
- Antigen name:DDB1 and CUL4 associated factor 6
- Antigen symbol:DCAF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:55827
- Antigen synonyms:MSTP055|IQWD1|PC326|ARCAP|NRIP|1200006M05Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: NRIP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: