Anti-UEV2 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56360
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2 Variant 2
- Antigen symbol:UEV2V2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:7336
- Antigen synonyms:ubiquitin-conjugating enzyme E2v2|S. cerevisiae|UEV2|DDVit-1|EDPF-1|DDVIT1|1 alpha,25-dihydroxyvitamin D3-inducible|UBE2V2|EDPF1|MMS2|ubiquitin-conjugating enzyme E2 variant 2|EDAF-1|UEV-2|methyl methanesulfonate sensitive 2|enterocyte differentiation promoting factor|homolog of
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: UEV2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human