Anti-Vitamin D Receptor Rabbit Polyclonal Antibody
Catalog # BSBTPA1983
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Vitamin D Receptor
- Antigen symbol:VDR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:vitamin D receptor|NR1I1|vitamin D3 receptor|1,25-dihydroxyvitamin D3 receptor
- Amino acid number:389 - 404
- Sequence:DLRSLNEEHSKQYRCL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Vitamin D Receptor (389-404aa DLRSLNEEHSKQYRCL), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Vitamin D3 receptor (VDR) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Vitamin D Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat