Anti-DDX4/MVH Rabbit Polyclonal Antibody
Catalog # BSBTPA1963
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:DDX4/MVH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:MVH
- Amino acid number:253 - 272
- Sequence:EDEDSIFAHYQTGINFDKYD
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human DDX4 (253-272aa EDEDSIFAHYQTGINFDKYD), identical to the the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Probable ATP-dependent RNA helicase DDX4 (DDX4) detection. Tested with WB, IHC-P, IHC-F, ICC in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: DDX4/MVH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat