Anti-LOX 1 Rabbit Polyclonal Antibody
BSBTPA1833
- Antibody type:Primary
- Antigen name:Oxidized Low Density Lipoprotein Receptor 1
- Antigen symbol:OLR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:SCARE1|CLEC8A|LOX1|SLOX1|LOXIN
- Amino acid number:114 - 127
- Sequence:NEKSKEQMELHHQN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Immunogen A synthetic peptide corresponding to a sequence in the middle region of human LOX 1 (114-127aa NEKSKEQMELHHQN), different from the related rat sequence by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Oxidized low-density lipoprotein receptor 1 (OLR1) detection. Tested with WB in Human; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: LOX 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat