Anti-Stra8 Rabbit Polyclonal Antibody
Catalog # BSBTPA1685
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Stimulated by retinoic acid 8
- Antigen symbol:STRA8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:313 - 330
- Sequence:DEEMIMLQCTETFDDEDL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Stra8 (313-330aa DEEMIMLQCTETFDDEDL).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Stimulated by retinoic acid gene 8 protein homolog (STRA8) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Stra8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human