Anti-Sialyl Tn Mouse Monoclonal Antibody [clone: STn 219]
Catalog # ABCAAB115957-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen symbol:Sialyl Tn
- Clonality:Monoclonal
- Clone:STn 219
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Sialyl Tn
Clonality: monoclonal
Clone: STn 219
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human