Anti-Trefoil Factor 3 Rabbit Monoclonal Antibody [clone: EPR3973]
ABCAAB109104-100
: Abcam
- Antibody type:Primary
- Antigen name:Trefoil factor 3
- Antigen symbol:TFF3
- Clonality:Monoclonal
- Clone:EPR3973
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: Trefoil Factor 3
Clonality: monoclonal
Clone: EPR3973
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: