- Antibody type:Primary
- Antigen name:Glutathione reductase
- Antigen symbol:GSR
- Clonality:Monoclonal
- Clone:AP-MAB0866
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2
- Reactivity:Mouse
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GSR (glutathione reductase)
Clonality: monoclonal
Clone: AP-MAB0866
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse