Order Entry
Ireland
ContactUsLinkComponent
Anti-GM-CSF Receptor alpha Mouse monoclonal antibody unconjugated [clone: 4H1]
Anti-GM-CSF Receptor alpha Mouse monoclonal antibody unconjugated [clone: 4H1]
Catalog # ABCAAB95681-100
Supplier:  Abcam
Anti-GM-CSF Receptor alpha Mouse monoclonal antibody unconjugated [clone: 4H1]
Catalog # ABCAAB95681-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GM-CSF Receptor alpha
  • Clonality:
    Monoclonal
  • Clone:
    4H1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GM-CSF Receptor alpha
Clonality: monoclonal
Clone: 4H1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human