Order Entry
Ireland
ContactUsLinkComponent
Anti-Macrophage inflammatory protein 5 Goat Polyclonal Antibody (Biotin)
Anti-Macrophage inflammatory protein 5 Goat Polyclonal Antibody (Biotin)
Catalog # ABCAAB83433-50
Supplier:  Abcam
Anti-Macrophage inflammatory protein 5 Goat Polyclonal Antibody (Biotin)
Catalog # ABCAAB83433-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Macrophage Inflammatory Protein 5
  • Antigen symbol:
    MIP5
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Macrophage inflammatory protein 5
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human