Order Entry
Ireland
ContactUsLinkComponent
Anti-Procollagen Type 1 Rat Monoclonal Antibody [clone: 2Q576]
Anti-Procollagen Type 1 Rat Monoclonal Antibody [clone: 2Q576]
Catalog # ABCAAB64409-50
Supplier:  Abcam
Anti-Procollagen Type 1 Rat Monoclonal Antibody [clone: 2Q576]
Catalog # ABCAAB64409-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Procollagen Type 1
  • Clonality:
    Monoclonal
  • Clone:
    2Q576
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Human
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Procollagen Type 1
Clonality: monoclonal
Clone: 2Q576
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Pig