Order Entry
Ireland
ContactUsLinkComponent
Anti-OATP2 Mouse Monoclonal Antibody [clone: MDQ]
Anti-OATP2 Mouse Monoclonal Antibody [clone: MDQ]
Catalog # ABCAAB15442-500
Supplier:  Abcam
Anti-OATP2 Mouse Monoclonal Antibody [clone: MDQ]
Catalog # ABCAAB15442-500
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    OATP2
  • Clonality:
    Monoclonal
  • Clone:
    MDQ
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µl
  • Pk:
    500 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: OATP2
Clonality: monoclonal
Clone: MDQ
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human