Anti-AMIGO1 Mouse Monoclonal Antibody
Catalog # BIRBORB99107-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:adhesion molecule with Ig-like domain 1
- Antigen symbol:AMIGO1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Amino acid number:554 to 574
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Fusion protein amino acids 554-574 (cytoplasmic C-terminus) of human AMIGO-1, accession number Q86WK6
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: AMIGO1 (adhesion molecule with Ig-like domain 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat