Anti-ACTG1 Mouse Monoclonal Antibody
BIRBORB98202-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:actin, gamma 1
- Antigen symbol:ACTG1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS containing 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: ACTG1 (actin, gamma 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: