Anti-ANK3 Mouse Monoclonal Antibody
Catalog # BIRBORB96368-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:ankyrin 3, Node Of Ranvier (ankyrin G)
- Antigen symbol:ANK3
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS pH 7,4, 50% glycerol and 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Fusion protein 1000 C-terminal amino acids of human Ankyrin G encompassing all of Ankyrin G with the exception of ankyrin repeats
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: ANK3 (ankyrin 3, node of Ranvier (ankyrin G))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: