Anti-NR2C2 Mouse Monoclonal Antibody
BIRBORB94775-50
: Biorbyt
- Antibody type:Primary
- Antigen name:nuclear receptor subfamily 2, group C, member 2
- Antigen symbol:NR2C2
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: NR2C2 (nuclear receptor subfamily 2, group C, member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human