Anti-ACOT9 Mouse Monoclonal Antibody
BIRBORB89783-50
: Biorbyt
- Antibody type:Primary
- Antigen name:acyl-CoA thioesterase 9
- Antigen symbol:ACOT9
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: ACOT9 (acyl-CoA thioesterase 9)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human