Anti-AG3 Mouse Monoclonal Antibody
BIRBORB89772-50
: Biorbyt
- Antibody type:Primary
- Antigen symbol:CD58
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:AG3
- Storage buffer:15 mM sodium azide, 0,2% BSA, PBS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 1 ?g/ml; Immunochemistry: 10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: AG3
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human