Anti-MCHR1 Rabbit Polyclonal Antibody
BIRBORB85477-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Melanin-Concentrating Hormone Receptor 1
- Antigen symbol:MCHR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 18 amino acid peptide from C-terminus of human MCHR1
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: MCHR1 (melanin-concentrating hormone receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human