Anti-Trichohyalin granules Mouse Monoclonal Antibody
Catalog # BIRBORB76830-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:Trichohyalin granules
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Storage buffer:Purified antibody containing PBS, 0,1% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Urea-mercaptoethanol extract of human hair follicles
- Purification:purified by Protein G chromatography
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Trichohyalin granules
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human