Anti-SYNCRIP Mouse Monoclonal Antibody
Catalog # BIRBORB76676-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:synaptotagmin binding, cytoplasmic RNA interacting Protein
- Antigen symbol:SYNCRIP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant untagged hnRNP-Q protein human
- Purification:purified by Protein A chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SYNCRIP (synaptotagmin binding, cytoplasmic RNA interacting protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human