Anti-SHC4 Rabbit Polyclonal Antibody
Catalog # BIRBORB69939-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:SHC Adaptor Protein 4
- Antigen symbol:SHC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Concentration:0,54 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SHC4 (SHC (Src homology 2 domain containing) family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human