Anti-Cytokeratin 8+18 Mouse monoclonal antibody
Catalog # BIRBORB137200-250
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 8/18
- Antigen symbol:CK8/CK18
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Environmentally Preferable:
- Format:Tissue culture supernatant
- Storage buffer:0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Cytokeratins purified from breast cancer MCF-7 cells.
- Purification:purified by affinity chromatography
- Size:250 µl
- Pk:250 µl
Specifications
About this item
Immunochemistry: 1:50
Type: Primary
Antigen: Cytokeratin 8+18
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human