Order Entry
Ireland
ContactUsLinkComponent
Anti-HK2 Rabbit Polyclonal Antibody
Anti-HK2 Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
undefined
Anti-HK2 Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB48965-200
  • BIRBORB48965-100
  • Antigen symbol:
  • HK2
  • HK2
  • Antigen name:
  • Hexokinase 2
  • Hexokinase 2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant protein of human HK2
  • Recombinant protein of human HK2
  • Molecular weight:
  • 102 kDa
  • 102 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Storage buffer:
  • Supplied as PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Supplied as PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together