Anti-CT1 Mouse monoclonal antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: CT1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human
Specifications
- Catalog No:
- BIRBORB48868-1000
- BIRBORB48868-200
- Antigen symbol:
- CT1
- CT1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Immunogen:
- Peptide
- Peptide
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together