Anti-GABA B receptor GB1a subunit Rabbit Polyclonal Antibody
BIRBORB44788-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:GABA B receptor GB1a subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: GABA B receptor GB1a subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat