Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Cyfra-21-1 Mouse Monoclonal Antibody
Catalog # BIRBORB25905-1
Supplier:  Biorbyt
Anti-Cyfra-21-1 Mouse Monoclonal Antibody
Catalog # BIRBORB25905-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Cyfra-21-1
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Soluble cytokeratin antigen fraction purified from human cell line.
  • Purification:
    purified by ion exchange chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Cyfra-21-1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human