Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Malondialdehyde (MDA) Goat Polyclonal Antibody
  BIRBORB25782-0.5
 :  Biorbyt
Anti-Malondialdehyde (MDA) Goat Polyclonal Antibody
  BIRBORB25782-0.5
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Malondialdehyde
  • Antigen symbol:
    MDA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage buffer:
    75 mM PBS, pH 7,2 containing 75 mM Sodium Chloride and 1 mM EDTA.
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml (OD280, E(0,1%)=1,35)
  • Shipping temperature:
    Ice
  • Immunogen:
    Malondialdehyde modified KLH
  • Purification:
    purified by MDA modified protein Sepharose affinity chromatography
  • Size:
    500 μg
  • Pk:
    0,5 mg

 

 

Western Blot: 1:200-1:20000

Type: Primary
Antigen: Malondialdehyde (MDA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: