Anti-LYPLAL1 Goat Polyclonal Antibody
Catalog # BIRBORB19782-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:lysophospholipase-like 1
- Antigen symbol:LYPLAL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Molecular weight:28 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:internal region
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:16000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: LYPLAL1 (lysophospholipase-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human