Anti-SLC39A5 Mouse Polyclonal Antibody
Catalog # ABNOH00283375-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 39 Member A5
- Antigen symbol:Solute carrier family 39, member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:283375
- Antigen synonyms:Zinc transporter ZIP5|Solute carrier family 39 member 5|LZT-Hs7|ZIP5|ZIP-5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: SLC39A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human