Anti-CNKSR3 Mouse Polyclonal Antibody
Catalog # ABNOH00154043-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CNKSR family member 3
- Antigen symbol:CNKSR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:154043
- Antigen synonyms:MAGI1
- Storage buffer:50 % glycerol
- Sequence:GGSSKCKQPLPGPKGSESPNSFLDQESRRRRFTIADSDQLPGYSVETNILPTKMREKTPSYGKPRPLSMPADGNWMGIVDPFARPRGHGRKGEDALCRY
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:CNKSR3 (NP_775786, 366 a.a. ~ 464 a.a) partial recombinant protein with GST tag.
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CNKSR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human