Anti-TWIST2 Mouse Monoclonal Antibody [clone: 3C12]
ABNOH00117581-M12
: Abnova
- Antibody type:Primary
- Antigen name:Twist-related protein 2
- Antigen symbol:TWIST2
- Clonality:Monoclonal
- Clone:3C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:117581
- Antigen synonyms:AMS|SETLSS|DERMO1|bHLHa39|FFDD3|BBRSAY
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TWIST2
Clonality: Monoclonal
Clone: 3C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human