Anti-UBTD2 Mouse Monoclonal Antibody [clone: 1B8-1B1]
ABNOH00092181-M01
: Abnova
- Antibody type:Primary
- Antigen name:Ubiquitin domain containing 2
- Antigen symbol:UBTD2
- Clonality:Monoclonal
- Clone:1B8-1B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:92181
- Antigen synonyms:DCUBP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: UBTD2
Clonality: Monoclonal
Clone: 1B8-1B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human