Anti-SLX4 (SLX4 structure-specific endonuclease Subunit) Mouse Polyclonal Antibody
Catalog # ABNOH00084464-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SLX4 structure-specific endonuclease Subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:84464
- Antigen synonyms:MUS312|FANCP|BTBD12
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: SLX4 (SLX4 structure-specific endonuclease Subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human