Anti-GPR61 Mouse Polyclonal Antibody
Catalog # ABNOH00083873-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 61
- Antigen symbol:GPR61
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:83873
- Antigen synonyms:BALGR|GPCR3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: GPR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human