Order Entry
Ireland
ContactUsLinkComponent
Anti-CDCA3 Mouse Monoclonal Antibody [clone: 4A10]
Anti-CDCA3 Mouse Monoclonal Antibody [clone: 4A10]
Catalog # ABNOH00083461-M10
Supplier:  Abnova
undefined
Anti-CDCA3 Mouse Monoclonal Antibody [clone: 4A10]
Catalog # ABNOH00083461-M10
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cell division cycle associated 3
  • Antigen symbol:
    CDCA3
  • Clonality:
    Monoclonal
  • Clone:
    4A10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    83461
  • Antigen synonyms:
    CDCA3|GRCC8|TOME-1|C8|Cell Division Cycle-associated protein 3|Gene-rich cluster protein C8|TOME1|Trigger of mitotic entry protein 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CDCA3
Clonality: Monoclonal
Clone: 4A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human