Order Entry
Ireland
ContactUsLinkComponent
Anti-EFHD1 Mouse Monoclonal Antibody [clone: 1A8]
Anti-EFHD1 Mouse Monoclonal Antibody [clone: 1A8]
  ABNOH00080303-M09
 :  Abnova
undefined
Anti-EFHD1 Mouse Monoclonal Antibody [clone: 1A8]
  ABNOH00080303-M09
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    EF-hand Domain Family, member D1
  • Antigen symbol:
    EFHD1
  • Clonality:
    Monoclonal
  • Clone:
    1A8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    80303
  • Antigen synonyms:
    MST133|MSTP133|EF-hand domain-containing protein D1|SWS2|PP3051|EF-hand domain-containing protein 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: EFHD1
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat