Anti-APOBEC3G Mouse Polyclonal Antibody
ABNOH00060489-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Apolipoprotein B mRNA Editing Enzyme, catalytic polypeptide 3G
- Antigen symbol:APOBEC3G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:60489
- Antigen synonyms:Apolipoprotein B mRNA Editing Enzyme|DNA dC->dU editing enzyme|bK150C2.7|ARP9|dJ494G10.1 |phorbolin-like protein MDS019|catalytic polypeptide-like 3G|CEM15|APOBEC3G|FLJ12740|OTTHUMP00000028911|MDS019|Apolipoprotein B mRNA Editing Enzyme catalytic polypeptide-like 3G
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: APOBEC3G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human