Order Entry
Ireland
ContactUsLinkComponent
Anti-SMYD2 Rabbit Polyclonal Antibody
Anti-SMYD2 Rabbit Polyclonal Antibody
Catalog # ABNOH00056950-D01P
Supplier:  Abnova
undefined
Anti-SMYD2 Rabbit Polyclonal Antibody
Catalog # ABNOH00056950-D01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SET and MYND domain containing 2
  • Antigen symbol:
    SMYD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    56950
  • Antigen synonyms:
    HSKM-B|KMT3C|SMYD2|SET and MYND domain-containing protein 2|Lysine N-methyltransferase 3C
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SMYD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human