Anti-PCDHA7 Mouse Monoclonal Antibody [clone: 4G3]
ABNOH00056141-M10
: Abnova
- Antibody type:Primary
- Antigen name:Protocadherin alpha 7
- Antigen symbol:PCDHA7
- Clonality:Monoclonal
- Clone:4G3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:56141
- Antigen synonyms:CNRN4|CRNR4|CNR4|CNRS4|PCDH-ALPHA7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PCDHA7
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human