Anti-UNC45A Mouse Polyclonal Antibody
Catalog # ABNOH00055898-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Unc-45 Homolog A
- Antigen symbol:UNC45A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55898
- Antigen synonyms:SMAP1|SMAP-1|UNC-45A|GC-UNC45|GCUNC45|IRO039700|GCUNC-45
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: UNC45A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human