Order Entry
Ireland
ContactUsLinkComponent
Anti-NCKIPSD Mouse Monoclonal Antibody [clone: 1A10]
Anti-NCKIPSD Mouse Monoclonal Antibody [clone: 1A10]
  ABNOH00051517-M11
 :  Abnova
undefined
Anti-NCKIPSD Mouse Monoclonal Antibody [clone: 1A10]
  ABNOH00051517-M11
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    NCK interacting protein with SH3 domain
  • Antigen symbol:
    NCKIPSD
  • Clonality:
    Monoclonal
  • Clone:
    1A10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    51517
  • Antigen synonyms:
    ORF1|WASLBP|AF3P21|VIP54|SPIN90|DIP1|DIP|WISH
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: NCKIPSD
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human